3-Ethyl-3-methylpentandisäure 95%

Lieferant: Apollo Scientific

OR936775-10G OR936775-1G OR936775-5G
APOSOR936775-10GEA 705 EUR
APOSOR936775-10G APOSOR936775-1G APOSOR936775-5G
3-Ethyl-3-methylpentandisäure 95%
3-Ethyl-3-methylpentandisäure
Formel: C₈H₁₄O₄
Molecular Weight: 174,20 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00020494
CAS-Nummer: 5345-01-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR