2-Methyl-4-phenylthiazol 95%

Lieferant: Apollo Scientific

Synonyme: 2-Methyl-4-phenylthiazole

OR936792-1G OR936792-5G
APOSOR936792-1GEA 87 EUR
APOSOR936792-1G APOSOR936792-5G
2-Methyl-4-phenylthiazol 95%
2-Methyl-4-phenylthiazol
Formel: C₁₀H₉NS
Molecular Weight: 175,25 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00489998
CAS-Nummer: 1826-16-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR