Benzyl isopropenyl ether 95%
Lieferant: Apollo Scientific
Synonyme:
2-Benzyloxy-1-propene, Isopropenyl Benzyl Ether
OR936797-1G
OR936797-250MG
APOSOR936797-1GEA
415
EUR
APOSOR936797-1G
APOSOR936797-250MG
Benzyl isopropenyl ether 95%
Benzyl isopropenyl ether
Formel:
C₁₀H₁₂O Molecular Weight: 148,21 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00059913 CAS-Nummer: 32783-20-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...