4-Chlor-(3-methoxycarbonyl)phenylboronsäure 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
4-Chloro-3-(methoxycarbonyl)benzeneboronic acid, 4-Chloro-3-(methoxycarbonyl)phenylboronic acid
OR9368-1G
OR9368-5G
OR9368-10G
APOSOR9368-1GEA
21.9
EUR
APOSOR9368-1G
APOSOR9368-5G
APOSOR9368-10G
4-Chlor-(3-methoxycarbonyl)phenylboronsäure 98%
4-Chlor-(3-methoxycarbonyl)phenylboronsäure
Formel:
C₈H₈BClO₄ Molecular Weight: 214,4107 g/mol Schmelzpunkt: 136…138 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD06801690 CAS-Nummer: 874219-45-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...