3-Chlor-2,4(3H,5H)-furandion 95%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
3-Chlorotetronic Acid, 3-Chlorofuran-2,4(3H,5H)-dione, 3-Chlorotetrahydrofuran-2,4-dione, 3-Chloro-4-hydroxy-2(5H)-furanone
OR936822-25G
OR936822-5G
APOSOR936822-25GEA
183
EUR
APOSOR936822-25G
APOSOR936822-5G
3-Chlor-2,4(3H,5H)-furandion 95%
3-Chlor-2,4(3H,5H)-furandion
Formel:
C₄H₃ClO₃ Molecular Weight: 134,52 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00134273 CAS-Nummer: 4971-55-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...