Methylthiosalicylat 95%
Lieferant: Apollo Scientific
Synonyme:
Methyl 2-mercaptobenzoate
OR936843-1G
OR936843-25G
OR936843-5G
OR936843-2.5G
OR936843-10G
APOSOR936843-1GEA
26.1
EUR
APOSOR936843-1G
APOSOR936843-25G
APOSOR936843-5G
APOSOR936843-2.5G
APOSOR936843-10G
Methylthiosalicylat 95%
Methylthiosalicylat
Formel:
C₈H₈O₂S Lagertemperatur: Raumtemperatur |
MDL:
MFCD00060678 CAS-Nummer: 4892-02-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...