2-[4-(Cyanmethyl)phenyl]acetamid 95%

Lieferant: Apollo Scientific

OR936870-1G OR936870-250MG
APOSOR936870-1GEA 763 EUR
APOSOR936870-1G APOSOR936870-250MG
2-[4-(Cyanmethyl)phenyl]acetamid 95%
2-[4-(Cyanomethyl)phenyl]acetamide
Formel: C₁₀H₁₀N₂O
Molecular Weight: 174,20 g/mol
Lagertemperatur: Raumtemperatur
CAS-Nummer: 99071-55-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR