2-sulphanyl-5,6,7,8-tetrahydro-4-quinazolinol

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-Hydroxy-5,6,7,8-tetrahydroquinazoline-2-thiol

OR9369-1G OR9369-5G
APOSOR9369-1GEA 140 EUR
APOSOR9369-1G APOSOR9369-5G
2-sulphanyl-5,6,7,8-tetrahydro-4-quinazolinol
2-sulphanyl-5,6,7,8-tetrahydro-4-quinazolinol
Formel: C₈H₁₀N₂OS
Molecular Weight: 182,25 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00665849
CAS-Nummer: 16064-21-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR