4-(Pentyloxy)phenol 95%
Lieferant: Apollo Scientific
Synonyme:
4-Pentyloxyphenol, Hydroquinone Monopentyl Ether, 4-(Pentyloxy)phenol, Hydroquinone Monoamyl Ether
OR936941-5G
APOSOR936941-5GEA
46.4
EUR
APOSOR936941-5G
4-(Pentyloxy)phenol 95%
4-(Pentyloxy)phenol
Formel:
C₁₁H₁₆O₂ Molecular Weight: 180,25 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00044283 CAS-Nummer: 18979-53-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...