2,2,2-Trimethylthioacetamide 95%

Lieferant: Apollo Scientific

Synonyme: 2,2-Dimethylpropanethioamide

OR936945-25G OR936945-5G
APOSOR936945-25GEA 278 EUR
APOSOR936945-25G APOSOR936945-5G
2,2,2-Trimethylthioacetamide 95%
2,2,2-Trimethylthioacetamide
Formel: C₅H₁₁NS
Molecular Weight: 117.21 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09742834
CAS-Nummer: 630-22-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR