3-Methoxybiphenyl 95%
Lieferant: Apollo Scientific
OR937041-10G
OR937041-1G
OR937041-5G
APOSOR937041-10GEA
812
EUR
APOSOR937041-10G
APOSOR937041-1G
APOSOR937041-5G
3-Methoxybiphenyl 95%
3-Methoxybiphenyl
Formel:
C₁₃H₁₂O Molecular Weight: 184.24 g/mol Siedepunkt: 140 °C/5.25mmHg Dichte: 1.08 g/cm³ (25 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00027514 CAS-Nummer: 2113-56-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...