1-Naphthylmalononitril 95%

Lieferant: Apollo Scientific

OR937052-1G OR937052-250MG
APOSOR937052-1GEA 734 EUR
APOSOR937052-1G APOSOR937052-250MG
1-Naphthylmalononitril 95%
1-Naphthylmalononitrile
Formel: C₁₃H₈N₂
Lagertemperatur: Raumtemperatur
MDL: MFCD00142672
CAS-Nummer: 5518-09-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR