4-Acetyl-N-methylaniline 95%

Lieferant: Apollo Scientific

Synonyme: 1-(4-(Methylamino)phenyl)ethanone, 4'-(Methylamino)acetophenone

OR937089-5G
APOSOR937089-5GEA 178 EUR
APOSOR937089-5G
4-Acetyl-N-methylaniline 95%
4-Acetyl-N-methylaniline
Formel: C₉H₁₁NO
Molecular Weight: 149,193 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09261160
CAS-Nummer: 17687-47-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR