Methyl-4-ethoxybenzoat 95%
Lieferant: Apollo Scientific
Synonyme:
4-Ethoxybenzoesäuremethylester
OR937109-100G
OR937109-25G
APOSOR937109-100GEA
340
EUR
APOSOR937109-100G
APOSOR937109-25G
Methyl-4-ethoxybenzoat 95%
Methyl-4-ethoxybenzoat
Formel:
C₁₀H₁₂O₃ Molecular Weight: 180,2 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00017264 CAS-Nummer: 23676-08-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...