2-Methylcyclohexanon 95%
Lieferant: Apollo Scientific
OR937162-100G
APOSOR937162-100GEA
183
EUR
APOSOR937162-100G
2-Methylcyclohexanon 95%
2-Methylcyclohexanone
Formel:
C₇H₁₂O Molecular Weight: 112,17 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00001635 CAS-Nummer: 583-60-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...