4-Ethylbenzonitril 95%

Lieferant: Apollo Scientific

Synonyme: p-Ethylbenzonitrile

OR937171-25G
APOSOR937171-25GEA 106 EUR
APOSOR937171-25G
4-Ethylbenzonitril 95%
4-Ethylbenzonitril
Formel: C₉H₉N
Molecular Weight: 131,18 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00016383
CAS-Nummer: 25309-65-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR