(R)-(+)-1-(2-Naphthyl)ethylamin 95%

Lieferant: Apollo Scientific

Synonyme: (R)-1-(2-Naphthyl)ethylamine

OR937174-5G
APOSOR937174-5GEA 65.4 EUR
APOSOR937174-5G
(R)-(+)-1-(2-Naphthyl)ethylamin 95%
(R)-(+)-1-(2-Naphthyl)ethylamin
Formel: C₁₂H₁₃N
Molecular Weight: 171.24 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00085367
CAS-Nummer: 3906-16-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR