N-Vinyl-ε-caprolactam 95%

Lieferant: Apollo Scientific

Synonyme: 1-Vinylazepan-2-one

OR937197-500G
APOSOR937197-500GEA 71.2 EUR
APOSOR937197-500G
N-Vinyl-ε-caprolactam 95%
N-Vinyl-ε-caprolactam
Formel: C₈H₁₃NO
Molecular Weight: 139,20 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00080693
CAS-Nummer: 2235-00-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR