3-Nonen-2-on 95%

Lieferant: Apollo Scientific

Synonyme: Non-3-en-2-one

OR937230-25G OR937230-100G
APOSOR937230-25GEA 72.5 EUR
APOSOR937230-25G APOSOR937230-100G
3-Nonen-2-on 95%
3-Nonen-2-on
Formel: C₉H₁₆O
Molecular Weight: 140,23 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00010241
CAS-Nummer: 14309-57-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR