6-tert-Butyl-1-indanone 95%
Lieferant: Apollo Scientific
Synonyme:
6-(tert-Butyl)-2,3-dihydro-1H-inden-1-one
OR937241-1G
OR937241-5G
APOSOR937241-1GEA
74.1
EUR
APOSOR937241-1G
APOSOR937241-5G
6-tert-Butyl-1-indanone 95%
6-tert-Butyl-1-indanone
Formel:
C₁₃H₁₆O Molecular Weight: 188.2655 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD11590864 CAS-Nummer: 162752-17-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...