4-(3-Buten-1-yloxy)benzoesäure 95%
Lieferant: Apollo Scientific
Synonyme:
4-(But-3-en-1-yloxy)benzoic acid
OR937299-5G
APOSOR937299-5GEA
68.3
EUR
APOSOR937299-5G
4-(3-Buten-1-yloxy)benzoesäure 95%
4-(3-Buten-1-yloxy)benzoesäure
Formel:
C₁₁H₁₂O₃ Molecular Weight: 192,21 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00191325 CAS-Nummer: 115595-27-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...