2-Amino-5,6-dimethylbenzothiazol 95%

Lieferant: Apollo Scientific

Synonyme: 5,6-Dimethylbenzo[d]thiazol-2-amine

OR937310-5G
APOSOR937310-5GEA 68.3 EUR
APOSOR937310-5G
2-Amino-5,6-dimethylbenzothiazol 95%
2-Amino-5,6-dimethylbenzothiazole
Formel: C₉H₁₀N₂S
Molecular Weight: 178,2500 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00005790
CAS-Nummer: 29927-08-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR