α-Chlorpropiophenon 95%

Lieferant: Apollo Scientific

Synonyme: 2-Chloro-1-phenyl-1-propanone, 2-Chloropropiophenone

OR937361-1G OR937361-25G OR937361-5G
APOSOR937361-1GEA 125 EUR
APOSOR937361-1G APOSOR937361-25G APOSOR937361-5G
α-Chlorpropiophenon 95%
α-Chlorpropiophenon
Formel: C₉H₉ClO
Lagertemperatur: Raumtemperatur
MDL: MFCD03844782
CAS-Nummer: 6084-17-9

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR