1-(Dichlormethoxy)butan 95%
Lieferant: Apollo Scientific
Synonyme:
Dichloromethyl Butyl Ether
OR937366-10G
OR937366-1G
OR937366-5G
APOSOR937366-10GEA
386
EUR
APOSOR937366-10G
APOSOR937366-1G
APOSOR937366-5G
1-(Dichlormethoxy)butan 95%
Butyl dichloromethyl ether
Formel:
C₅H₁₀Cl₂O Molecular Weight: 157,03 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00191420 CAS-Nummer: 5312-73-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...