(S)-(-)-1-Ethyl-2-Pyrrolidincarboxylsäure-Ethylester 95%

Lieferant: Apollo Scientific

Synonyme: (-)-1-Ethyl-L-proline Ethyl Ester, (S)-(-)-1-Ethyl-2-pyrrolidinecarboxylic Acid Ethyl Ester, Et-Pro-OEt

OR937412-1G OR937412-5G
APOSOR937412-1GEA 203 EUR
APOSOR937412-1G APOSOR937412-5G
(S)-(-)-1-Ethyl-2-Pyrrolidincarboxylsäure-Ethylester 95%
Ethyl (S)-(-)-1-ethyl-2-pyrrolidinecarboxylate
Formel: C₉H₁₇NO₂
Molecular Weight: 171.24 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00191434
CAS-Nummer: 938-54-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR