4-Isopropylpiperidin 95%

Lieferant: Apollo Scientific

OR937445-1G OR937445-250MG
APOSOR937445-1GEA 393 EUR
APOSOR937445-1G APOSOR937445-250MG
4-Isopropylpiperidin 95%
4-Isopropylpiperidin
Formel: C₈H₁₇N
Lagertemperatur: Raumtemperatur
MDL: MFCD09055342
CAS-Nummer: 19678-58-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR