N- (2-Methoxyethyl) Isopropylamin 95%

Lieferant: Apollo Scientific

OR937475-1G OR937475-5G
APOSOR937475-1GEA 39.3 EUR
APOSOR937475-1G APOSOR937475-5G
N- (2-Methoxyethyl) Isopropylamin 95%
N- (2-Methoxyethyl) Isopropylamin
Formel: C₆H₁₅NO
Molecular Weight: 117,19 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00144828
CAS-Nummer: 104678-18-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR