Methacrylsäure N-Hydroxysuccinimid-Ester 95%

Lieferant: Apollo Scientific

Synonyme: N-Methacryloyloxysuccinimide, Methacrylic Acid N-Succinimidyl Ester

OR937503-5G
APOSOR937503-5GEA 142 EUR
APOSOR937503-5G
Methacrylsäure N-Hydroxysuccinimid-Ester 95%
Methacrylsäure N-Hydroxysuccinimid-Ester
Formel: C₈H₉NO₄
Molecular Weight: 183,16 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD15072193
CAS-Nummer: 38862-25-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR