6-Brom-1H-[1,2,3]triazolo[4,5-b]pyridin 98%
Lieferant: Apollo Scientific
OR937506-1G
OR937506-250MG
OR937506-5G
APOSOR937506-1GEA
66.7
EUR
APOSOR937506-1G
APOSOR937506-250MG
APOSOR937506-5G
6-Brom-1H-[1,2,3]triazolo[4,5-b]pyridin 98%
6-Brom-1H-[1,2,3]triazolo[4,5-b]pyridin
Formel:
C₅H₃BrN₄ Lagertemperatur: Raumtemperatur |
MDL:
MFCD04060716 CAS-Nummer: 92276-38-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...