4-Isothiocyanato-1-buten 95%

Lieferant: Apollo Scientific

Synonyme: Isothiocyanic Acid 3-Buten-1-yl Ester

OR937510-1G OR937510-250MG
APOSOR937510-1GEA 604 EUR
APOSOR937510-1G APOSOR937510-250MG
4-Isothiocyanato-1-buten 95%
4-Isothiocyanato-1-buten
Formel: C₅H₇NS
Molecular Weight: 113.18 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00191660
CAS-Nummer: 3386-97-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR