1-(3-Brompropyl)-1H-pyrrol 95%

Lieferant: Apollo Scientific

OR937517-1G OR937517-250MG
APOSOR937517-1GEA 249 EUR
APOSOR937517-1G APOSOR937517-250MG
1-(3-Brompropyl)-1H-pyrrol 95%
1-(3-Bromopropyl)pyrrole
Formel: C₇H₁₀BrN
Molecular Weight: 188.07 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00191324
CAS-Nummer: 100779-91-7

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR