4-Octanol 97%
Lieferant: Apollo Scientific
Synonyme:
4-Hydroxyoctan, Propyl-butylcarbinol, Butyl-propylcarbinol
OR937548-25G
OR937548-5G
APOSOR937548-25GEA
478
EUR
APOSOR937548-25G
APOSOR937548-5G
4-Octanol 97%
4-Octanol
Formel:
C₈H₁₈O Molecular Weight: 130,23 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00014409 CAS-Nummer: 589-62-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...