(4-Chlorphenyl)acetylchlorid 97%

Lieferant: Apollo Scientific

Synonyme: (4-Chlorphenyl)essigsäurechlorid

OR937565-100G
APOSOR937565-100GEA 98.6 EUR
APOSOR937565-100G
(4-Chlorphenyl)acetylchlorid 97%
(4-Chlorphenyl)acetylchlorid
Formel: (C₆H₄Cl)CH₂COCl
Molecular Weight: 189,04 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00037111
CAS-Nummer: 25026-34-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR