2,8-Dichlorcyclooctanon 95%
Lieferant: Apollo Scientific
OR937597-1G
OR937597-250MG
APOSOR937597-1GEA
464
EUR
APOSOR937597-1G
APOSOR937597-250MG
2,8-Dichlorcyclooctanon 95%
2,8-Dichlorocyclooctanone
Formel:
C₈H₁₂Cl₂O Molecular Weight: 195,08 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD06797089 CAS-Nummer: 108249-93-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...