3-Methyl-1-hexanol 95%

Lieferant: Apollo Scientific

OR937638-1G
APOSOR937638-1GEA 119 EUR
APOSOR937638-1G
3-Methyl-1-hexanol 95%
3-Methyl-1-hexanol
Formel: C₇H₁₆O
Lagertemperatur: Raumtemperatur
MDL: MFCD01321178
CAS-Nummer: 13231-81-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR