4-Ethoxy-3-methoxybenzylalkohol 97%

Lieferant: Apollo Scientific

OR937675-1G OR937675-250MG
APOSOR937675-1GEA 0 EUR
APOSOR937675-1G APOSOR937675-250MG
4-Ethoxy-3-methoxybenzylalkohol 97%
4-Ethoxy-3-methoxybenzylalkohol
Formel: C₁₀H₁₄O₃
Lagertemperatur: Raumtemperatur
MDL: MFCD00004656
CAS-Nummer: 61813-58-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR