2,4-Diethyl-6-methyl-1,3,5-trioxane 95%

Lieferant: Apollo Scientific

OR937681-1G OR937681-250MG
APOSOR937681-1GEA 0 EUR
APOSOR937681-1G APOSOR937681-250MG
2,4-Diethyl-6-methyl-1,3,5-trioxane 95%
2,4-Diethyl-6-methyl-1,3,5-trioxane
Formel: C₈H₁₆O₃
Lagertemperatur: Raumtemperatur
MDL: MFCD00059793
CAS-Nummer: 117888-04-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR