N-Methylfurohydroxamsäure 95%
Lieferant: Apollo Scientific
Synonyme:
N-MFHA, N-(2-Furoyl)-N-methylhydroxylamine
OR937684-1G
OR937684-250MG
APOSOR937684-1GEA
464
EUR
APOSOR937684-1G
APOSOR937684-250MG
N-Methylfurohydroxamsäure 95%
N-Methylfurohydroxamsäure
Formel:
C₆H₇NO₃ Molecular Weight: 141,13 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00144939 CAS-Nummer: 109531-96-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...