N-(Diethylcarbamoyl)-N-Methoxyformamid 95%
Lieferant: Apollo Scientific
Synonyme:
N,N-Diethyl-N'-formyl-N'-methoxyurea
OR937688-1G
OR937688-250MG
APOSOR937688-1GEA
819
EUR
APOSOR937688-1G
APOSOR937688-250MG
N-(Diethylcarbamoyl)-N-Methoxyformamid 95%
N-(Diethylcarbamoyl)-N-Methoxyformamid
Formel:
C₇H₁₄N₂O₃ Molecular Weight: 174.20 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00191345 CAS-Nummer: 146039-03-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...