2-Methyl-6-nitrobenzoyl chloride 95%
Lieferant: Apollo Scientific
Synonyme:
6-Nitro-o-toluoyl Chloride
OR937743-1G
OR937743-5G
APOSOR937743-1GEA
255
EUR
APOSOR937743-1G
APOSOR937743-5G
2-Methyl-6-nitrobenzoyl chloride 95%
2-Methyl-6-nitrobenzoyl chloride
Formel:
C₈H₆ClNO₃ Molecular Weight: 199,59 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD06797116 CAS-Nummer: 66232-57-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...