9-Methyl-3,4-dihydro-2H-pyrido[1,2-a]pyrimidin-2-on 95%

Lieferant: Apollo Scientific

Synonyme: Acid Captor 9M

OR937763-25G OR937763-5G
APOSOR937763-25GEA 357 EUR
APOSOR937763-25G APOSOR937763-5G
9-Methyl-3,4-dihydro-2H-pyrido[1,2-a]pyrimidin-2-on 95%
9-Methyl-3,4-dihydro-2H-pyrido[1,2-a]pyrimidin-2-on
Formel: C₉H₁₀N₂O
Molecular Weight: 162.19 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00059749
CAS-Nummer: 61751-44-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR