Ethyl-2-hydroxybutyrate 95%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
Ethyl 2-Hydroxybutyrate, 2-Hydroxybutyric Acid Ethyl Ester, Ethyl 2-hydroxybutanoate
OR937766-1G
OR937766-25G
OR937766-5G
APOSOR937766-1GEA
55.1
EUR
APOSOR937766-1G
APOSOR937766-25G
APOSOR937766-5G
Ethyl-2-hydroxybutyrate 95%
Ethyl-2-hydroxybutyrate
Formel:
C₆H₁₂O₃ Molecular Weight: 132,16 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00014407 CAS-Nummer: 52089-54-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...