[(2-Chlorethoxy)methyl]benzol 98%
Lieferant: Apollo Scientific
Synonyme:
2-(Benzyloxy)ethyl Chloride, ((2-Chloroethoxy)methyl)benzene, (2-Chloroethoxymethyl)benzene
OR937793-25G
APOSOR937793-25GEA
59.6
EUR
APOSOR937793-25G
[(2-Chlorethoxy)methyl]benzol 98%
[(2-Chlorethoxy)methyl]benzol
Formel:
C₉H₁₁ClO Molecular Weight: 170,64 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00018964 CAS-Nummer: 17229-17-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...