2,2,5,6-Tetramethyl-4H-1,3-dioxin-4-on 95%
Lieferant: Apollo Scientific
OR937815-10G
OR937815-1G
OR937815-5G
APOSOR937815-10GEA
719
EUR
APOSOR937815-10G
APOSOR937815-1G
APOSOR937815-5G
2,2,5,6-Tetramethyl-4H-1,3-dioxin-4-on 95%
2,2,5,6-Tetramethyl-4H-1,3-dioxin-4-one
Formel:
C₈H₁₂O₃ Molecular Weight: 156,18 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00060139 CAS-Nummer: 87769-39-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...