2-Methoxy-1-butanol 95%

Lieferant: Apollo Scientific

OR937857-10G OR937857-1G OR937857-5G
APOSOR937857-10GEA 647 EUR
APOSOR937857-10G APOSOR937857-1G APOSOR937857-5G
2-Methoxy-1-butanol 95%
2-Methoxy-1-butanol
Formel: C₅H₁₂O₂
Molecular Weight: 104.15 g/mol
Lagertemperatur: Raumtemperatur
CAS-Nummer: 15467-25-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR