(2-Chlorpropoxy)benzol 95%

Lieferant: Apollo Scientific

Synonyme: 2-Chloropropyl Phenyl Ether

OR937869-10G OR937869-1G OR937869-5G
APOSOR937869-10GEA 734 EUR
APOSOR937869-10G APOSOR937869-1G APOSOR937869-5G
(2-Chlorpropoxy)benzol 95%
1-Phenoxy-2-chloropropane
Formel: C₉H₁₁ClO
Molecular Weight: 170,64 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00059488
CAS-Nummer: 53491-30-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR