2-Methylcyclohexyl-acetat 95%

Lieferant: Apollo Scientific

Synonyme: Acetic Acid 2-Methylcyclohexyl Ester

OR937875-25G OR937875-5G
APOSOR937875-25GEA 197 EUR
APOSOR937875-25G APOSOR937875-5G
2-Methylcyclohexyl-acetat 95%
2-Methylcyclohexyl acetate
Formel: C₉H₁₆O₂
Molecular Weight: 156,23 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00021390
CAS-Nummer: 5726-19-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR