(2S)-2-(Isopropylamino)-3-methyl-1-butanol 95%

Lieferant: Apollo Scientific

Synonyme: (S)-N-Isopropyl-2-(3-methyl-1-butanol)amine

OR937893-1G
APOSOR937893-1GEA 171 EUR
APOSOR937893-1G
(2S)-2-(Isopropylamino)-3-methyl-1-butanol 95%
(S)-2-Isopropylamino-3-methyl-1-butanol
Formel: C₈H₁₉NO
Molecular Weight: 145,25 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00671550
CAS-Nummer: 112211-88-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR