1,3-Dichlor-2-(methoxymethoxy)propan 95%

Lieferant: Apollo Scientific

Synonyme: 1,3-Dichloro-2-(methoxymethoxy)propane, 1,1-Bis(chloromethyl)-2,4-dioxapentane

OR937921-1G OR937921-25G OR937921-5G
APOSOR937921-1GEA 56.7 EUR
APOSOR937921-1G APOSOR937921-25G APOSOR937921-5G
1,3-Dichlor-2-(methoxymethoxy)propan 95%
1-Chloro-2-(chloromethyl)-3,5-dioxahexane
Formel: C₅H₁₀Cl₂O₂
Molecular Weight: 173.03 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00130114
CAS-Nummer: 70905-45-2

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR