Methyl-2,5-dihydro-3-thiophencarboxylat 95%
Lieferant: Apollo Scientific
Synonyme:
Methyl 2,5-dihydrothiophene-3-carboxylate, 2,5-Dihydrothiophene-3-carboxylic Acid Methyl Ester
OR937937-1G
OR937937-5G
APOSOR937937-1GEA
125
EUR
APOSOR937937-1G
APOSOR937937-5G
Methyl-2,5-dihydro-3-thiophencarboxylat 95%
Methyl-2,5-dihydrothiophene-3-carboxylate
Formel:
C₆H₈O₂S Molecular Weight: 144.19 g/mol Schmelzpunkt: 36 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD04038417 CAS-Nummer: 67488-46-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...